Anti-mCherry Rabbit Monoclonal Antibody [clone: EPR20579]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB232341-100 AB232341-1
ABCAAB232341-100EA 7600 DKK
ABCAAB232341-100 ABCAAB232341-1
Anti-mCherry Rabbit Monoclonal Antibody [clone: EPR20579]
Antistoffer
Rabbit monoclonal [EPR20579] to mCherry - BSA and Azide free.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: mCherry
Clonality: Monoclonal
Clone: EPR20579
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR