Anti-LIN7A Rabbit Monoclonal Antibody [clone: EPR12609]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB249802-100 AB249802-1
ABCAAB249802-100EA 7600 DKK
ABCAAB249802-100 ABCAAB249802-1
Anti-LIN7A Rabbit Monoclonal Antibody [clone: EPR12609]
Antistoffer
Rabbit monoclonal [EPR12609] to LIN7A - BSA and Azide free.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LIN7A
Clonality: Monoclonal
Clone: EPR12609
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR