Anti-p40 - DeltaNp63 Rabbit Monoclonal Antibody [clone: EPR17863-47]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB203826-100 AB203826-1ML
ABCAAB203826-100EA 5880 DKK
ABCAAB203826-100 ABCAAB203826-1ML
Anti-p40 - DeltaNp63 Rabbit Monoclonal Antibody [clone: EPR17863-47]
Antistoffer
Rabbit monoclonal [EPR17863-47] to p40 - DeltaNp63.
Type: Primary
Antigen: p40 - DeltaNp63
Clonality: Monoclonal
Clone: EPR17863-47
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat