Anti-Ubiquitin Mouse Monoclonal Antibody [clone: P4D1]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB303665-100 AB303665-1
ABCAAB303665-100EA 7600 DKK
ABCAAB303665-100 ABCAAB303665-1
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: P4D1]
Antistoffer
Mouse monoclonal [P4D1] to Ubiquitin - BSA and Azide free.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: P4D1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human; Mouse; Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR