Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR22076]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB243228-100 AB243228-20UL AB243228-1ML
ABCAAB243228-100EA 5320 DKK
ABCAAB243228-100 ABCAAB243228-20UL ABCAAB243228-1ML
Anti-CD28 Rabbit Monoclonal Antibody [clone: EPR22076]
Antistoffer
Rabbit monoclonal [EPR22076] to CD28.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: EPR22076
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR