Anti-PspCas13b Mouse Monoclonal Antibody [clone: 12F2-A10]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB273440-100 AB273440-1MG AB273440-20UG
ABCAAB273440-100EA 4950 DKK
ABCAAB273440-100 ABCAAB273440-1MG ABCAAB273440-20UG
Anti-PspCas13b Mouse Monoclonal Antibody [clone: 12F2-A10]
Antistoffer
Mouse monoclonal [12F2-A10] to PspCas13b.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PspCas13b
Clonality: Monoclonal
Clone: 12F2-A10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR