Anti-Ubiquitin (Linkage-specific K27) Rabbit Monoclonal Antibody [clone: EPR17034]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB238442-100 AB238442-1
ABCAAB238442-100EA 7600 DKK
ABCAAB238442-100 ABCAAB238442-1
Anti-Ubiquitin (Linkage-specific K27) Rabbit Monoclonal Antibody [clone: EPR17034]
Antistoffer
Rabbit monoclonal [EPR17034] to Ubiquitin (linkage-specific K27) - BSA and Azide free.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: EPR17034
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human; Recombinant full length protein