Anti-hnRNP U/p120 Rabbit Monoclonal Antibody [clone: EPR12278(2)(B)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB250283-1 AB250283-100
ABCAAB250283-1EA 68310 DKK
ABCAAB250283-1 ABCAAB250283-100
Anti-hnRNP U/p120 Rabbit Monoclonal Antibody [clone: EPR12278(2)(B)]
Antistoffer
Rabbit monoclonal [EPR12278(2)(B)] to hnRNP U/p120 - BSA and Azide free.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: hnRNP U/p120
Clonality: Monoclonal
Clone: EPR12278(2)(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR