Anti-S1P1/EDG1 Rabbit Monoclonal Antibody [clone: EPR21202]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB233386-100 AB233386-1ML
ABCAAB233386-100EA 5410 DKK
ABCAAB233386-100 ABCAAB233386-1ML
Anti-S1P1/EDG1 Rabbit Monoclonal Antibody [clone: EPR21202]
Antistoffer
Rabbit monoclonal [EPR21202] to S1P1/EDG1.

Type: Primary
Antigen: S1P1/EDG1
Clonality: Monoclonal
Clone: EPR21202
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR