Anti-PRDM1/Blimp1 Rat Monoclonal Antibody [clone: 6D3]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB252242-100 AB252242-1
ABCAAB252242-100EA 7600 DKK
ABCAAB252242-100 ABCAAB252242-1
Anti-PRDM1/Blimp1 Rat Monoclonal Antibody [clone: 6D3]
Antistoffer
Rat monoclonal [6D3] to PRDM1/Blimp1 - BSA and Azide free.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PRDM1/Blimp1
Clonality: Monoclonal
Clone: 6D3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR