Anti-Histone H3 (mono methyl K14) Rabbit Monoclonal Antibody [clone: EPR17708-235] (Alexa Fluor® 488)
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB307618-100 AB307618-10
ABCAAB307618-100EA 6110 DKK
ABCAAB307618-100 ABCAAB307618-10
Anti-Histone H3 (mono methyl K14) Rabbit Monoclonal Antibody [clone: EPR17708-235] (Alexa Fluor® 488)
Antistoffer
Alexa Fluor® 488 Rabbit monoclonal [EPR17708-235] to Histone H3 (mono methyl K14).
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR17708-235
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human