Anti-CDKN2A/p16INK4A +CDKN2B/p15INK4B Rabbit Monoclonal Antibody [clone: EP435Y-129R]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB219723-100 AB219723-1
ABCAAB219723-100EA 7600 DKK
ABCAAB219723-100 ABCAAB219723-1
Anti-CDKN2A/p16INK4A +CDKN2B/p15INK4B Rabbit Monoclonal Antibody [clone: EP435Y-129R]
Antistoffer
Rabbit monoclonal [EP435Y-129R] to CDKN2A/p16INK4A +CDKN2B/p15INK4B - BSA and Azide free.
Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CDKN2A/p16INK4A +CDKN2B/p15INK4B
Clonality: Monoclonal
Clone: EP435Y-129R
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human