Anti-Cytokeratin 3/CK-3 Mouse Monoclonal Antibody [clone: AE5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB68260-100 AB68260-1MG
ABCAAB68260-100EA 4900 DKK
ABCAAB68260-100 ABCAAB68260-1MG
Anti-Cytokeratin 3/CK-3 Mouse Monoclonal Antibody [clone: AE5]
Antistoffer
Mouse monoclonal [AE5] to Cytokeratin 3/CK-3.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cytokeratin 3/CK-3
Clonality: Monoclonal
Clone: AE5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR