Anti-Tartrate Resistant Acid Phosphatase Rabbit Monoclonal Antibody [clone: EPR15556]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB240970-100 AB240970-1
ABCAAB240970-100EA 7600 DKK
ABCAAB240970-100 ABCAAB240970-1
Anti-Tartrate Resistant Acid Phosphatase Rabbit Monoclonal Antibody [clone: EPR15556]
Antistoffer
Rabbit monoclonal [EPR15556] to Tartrate Resistant Acid Phosphatase - BSA and Azide free.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Tartrate Resistant Acid Phosphatase
Clonality: Monoclonal
Clone: EPR15556
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat