Anti-Ubiquitin Rabbit Monoclonal Antibody [clone: EPR17034]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB181537-100 AB181537-20UL AB181537-1ML
ABCAAB181537-100EA 5410 DKK
ABCAAB181537-100 ABCAAB181537-20UL ABCAAB181537-1ML
Anti-Ubiquitin Rabbit Monoclonal Antibody [clone: EPR17034]
Antistoffer
Rabbit monoclonal [EPR17034] to Ubiquitin (linkage-specific K27).

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Ubiquitin
Clonality: Monoclonal
Clone: EPR17034
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR