Anti-CD133 Rabbit Recombinant Multiclonal Antibody [clone: RM1029]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB284389-100 AB284389-20UL AB284389-1ML
ABCAAB284389-100EA 4530 DKK
ABCAAB284389-100 ABCAAB284389-20UL ABCAAB284389-1ML
Anti-CD133 Rabbit Recombinant Multiclonal Antibody [clone: RM1029]
Antistoffer
Rabbit recombinant multiclonal [RM1029] to CD133.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD133
Clonality: Recombinant Multiclonal
Clone: RM1029
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR