Anti-Apolipoprotein A I Rabbit Monoclonal Antibody [clone: EPR25446-111]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB307089-100 AB307089-20UL AB307089-1ML
ABCAAB307089-100EA 5180 DKK
ABCAAB307089-100 ABCAAB307089-20UL ABCAAB307089-1ML
Anti-Apolipoprotein A I Rabbit Monoclonal Antibody [clone: EPR25446-111]
Antistoffer
Rabbit monoclonal [EPR25446-111] to Apolipoprotein A I.
Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Apolipoprotein A I
Clonality: Monoclonal
Clone: EPR25446-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse