Anti-Coronin 1a/TACO Rabbit Monoclonal Antibody [clone: EPR19467-36]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB203698-100 AB203698-1ML
ABCAAB203698-100EA 5410 DKK
ABCAAB203698-100 ABCAAB203698-1ML
Anti-Coronin 1a/TACO Rabbit Monoclonal Antibody [clone: EPR19467-36]
Antistoffer
Rabbit monoclonal [EPR19467-36] to Coronin 1a/TACO.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Coronin 1a/TACO
Clonality: Monoclonal
Clone: EPR19467-36
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat