Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC98]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB240360-100 AB240360-1
ABCAAB240360-100EA 7600 DKK
ABCAAB240360-100 ABCAAB240360-1
Anti-beta Amyloid 1-42 Rabbit Monoclonal Antibody [clone: mOC98]
Antistoffer
Rabbit monoclonal [mOC98] to beta Amyloid 1-42 - BSA and Azide free.
Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: beta Amyloid 1-42
Clonality: Monoclonal
Clone: mOC98
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse