Anti-STING Rabbit Monoclonal Antibody [clone: SP339]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

AB227705-1 AB227705-100 AB227705-500 AB227705-20UL
ABCAAB227705-1EA 51950 DKK
ABCAAB227705-1 ABCAAB227705-100 ABCAAB227705-500 ABCAAB227705-20UL
Anti-STING Rabbit Monoclonal Antibody [clone: SP339]
Antistoffer
Rabbit monoclonal [SP339] to STING.

Advarsel: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: STING
Clonality: Monoclonal
Clone: SP339
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR