Cell culture dishes with Cell+ surface, Sarstedt SUREGrip design
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
83.1801.003 83.1800.003 83.1802.003
SARS83.1801.003EA 0 DKK
SARS83.1801.003 SARS83.1800.003 SARS83.1802.003
Cell culture dishes with Cell+ surface, Sarstedt SUREGrip design
Dyrkningsplader Celledyrkningsskåle
PS. Cell+ TC dishes for the growth of primary cultures and other fastidious cell lines. Convenient handling due to a raised ring around the circumference of the dish base, enabling an easy and secure grip of both the dish and the lid, even from a whole stack.