Cell culture dishes for suspension and hybridoma cells, Sarstedt SUREGrip design
Leverandør: SARSTEDT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
83.1800.002 83.1801.002 83.3902.500
SARS83.1800.002EA 0 DKK
SARS83.1800.002 SARS83.1801.002 SARS83.3902.500
Cell culture dishes for suspension and hybridoma cells, Sarstedt SUREGrip design
Dyrkningsplader Celledyrkningsskåle
TC treated dishes with convenient handling due to a raised ring around the circumference of the dish base, enabling an easy and secure grip of both the dish and the lid, even from a whole stack.