Petri dishes, square
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
688102_240 688161_240
391-2080EA 1650 DKK
391-2080 391-0465
Petri dishes, square
Dyrkningsplader Petriskåle
High grade PS. Square Petri dishes with exceptional optical clarity for microscopic analysis as well as heat resistance for use with hot agar.