Calibration weights, OIML, class F2
Leverandør: Sartorius Balances
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
YCW5548-00 YCW6148-00 YCW5148-02 YCW5248-00 YCW6248-00 YCW6148-02 YCW6548-00
SARBYCW5548-00EA 1900 DKK
SARBYCW5548-00 SARBYCW6148-00 SARBYCW5148-02 SARBYCW5248-00 SARBYCW6248-00 SARBYCW6148-02 SARBYCW6548-00
Calibration weights, OIML, class F2
Lodder Kalibreringslodder
These F2 class individual weights are from 100 g to 10 kg and made of non-magnetisable, stainless steel.
  • Cylindrical in shape
  • Density of 8,0 g/dm³
  • Highly polished
  • Highly corrosion-resistant
Leveringsinformation: Up to 1 kg supplied in plastic case and from 5 kg in a wooden case.