Empty solid phase extraction cartridges, Bond Elut™
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12131007 12131008 12131009 12131010 12131011 12131012
551-0356EA 1410 DKK
551-0356 551-0357 551-0358 551-0359 551-0360 551-0361
Empty solid phase extraction cartridges, Bond Elut™
Kromatografikolonner
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
  • Uniform batch-to-batch size for consistent performance
  • Cartridges are available from 1 to 60 ml
Bestillingsoplysninger: Frits has to be ordered separately