Filtering apparatus, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
257105451 257106304
516-0431EA 6340 DKK
516-0431 SCOT257106304
Filtering apparatus, DURAN®
Filtre Membranfiltertragte
Filtration apparatus with unit made from clear DURAN® borosilicate glass 3.3, PTFE insert and anodised aluminium clamp.
  • Scaled funnel
  • All components are available as spare parts
Typical applications: Coarse and ultrafine filtration, filtration of HPLC media, residue analysis. Filter paper, membrane filters (47 mm) or glass filters can be used for filtration.