Tubing clamps, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KT4.5 KT6 KT14 KT10
229-0540EA 409 DKK
229-0540 229-0541 BIBBKT14 BIBBKT10
Tubing clamps, Quickfit®
Klemmer Rørklemmer
Tubing clamps made of thermoplastic polyester, for use with flexible rubber and plastic tubing.
  • Provides fine flow control of liquids and gases
  • Easy one-handed adjustment: just roll serrated wheel along sloped floor