Joint clips for spherical joints, POM, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
KS13 KS19 KS29 KS35
201-8179EA 287 DKK
201-8179 201-8180 201-8181 BIBBKS35
Joint clips for spherical joints, POM, Quickfit®
Klemmer Klemmer til NS-slib
Clips made of polyacetal (POM) for retaining glassware having Quickfit® spherical joints.
  • Easy to fit and remove
  • Can be used at temperatures up to 80 °C
  • Clips are colour-coded for easy identification