Capillary columns, LIPODEX®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
723360.25 723360.50 723362.25 723362.50 723364.25 723364.50 723366.25 723366.50 723368.25 723368.50 723379.25 723379.50 723382.10
MANA723360.25EA 19200 DKK
MANA723360.25 MANA723360.50 MANA723362.25 MANA723362.50 MANA723364.25 MANA723364.50 MANA723366.25 MANA723366.50 MANA723368.25 MANA723368.50 MANA723379.25 MANA723379.50 MANA723382.10
Capillary columns, LIPODEX®
Kromatografikolonner
Cyclodextrin phases for GC enantiomer separation.
LIPODEX® A is recommended for carbohydrates, polyols, diols, hydroxycarboxylic acid esters, (epoxy‑) alcohols, glycerol derivatives, spiroacetals, ketones, alkyl halides.
Phase: Hexakis-(2,3,6-tri-O-pentyl)-α-cyclodextrin
LIPODEX® B is ideal for lactones, diols (cyclic carbonates), aminols, aldols (O-TFA), glycerol derivatives (cyclic carbonates).
Phase: Hexakis-(2,6-di-O-pentyl-3-O-acetyl)-α-cyclodextrin
LIPODEX® C is recommended for alcohols, cyanhydrins, olefins, hydroxycarboxylic acid esters, alkyl halides.
Phase: Heptakis-(2,3,6-tri-O-pentyl)-β-cyclodextrin
LIPODEX® D is recommended for amines (TFA), aminols (TFA), trans-cycloalkane-1,2-diols, trans-cycloalkane-1,3-diols (TFA), β-amino acid esters.
Phase: Heptakis-(2,6-di-O-pentyl-3-O-acetyl)-β-cyclodextrin
LIPODEX® E is ideal for α-amino acids, α- and β-hydroxycarboxylic acid esters, alcohols (TFA), diols (TFA), ketones, pheromones (cyclic acetals), amines, alkyl halides, lactones.
Phase: Octakis-(2,6-di-O-pentyl-3-O-butyryl)-γ-cyclodextrin
LIPODEX® G is recommended for menthol isomers, ketones, alcohols, carboxylic acid esters, terpenes.
Phase: Octakis-(2,3-di-O-pentyl-6-O-methyl)-γ-cyclodextrin
Max. temperature for isothermal operation 200 °C, max. temperature for short isotherms in a temperature program 220 °C.
A large number of separations have been achieved and published in the literature. However, it is not possible to make a general prediction, which phase could solve a given separation task.