HPLC columns, Pursuit™ UPS1,9
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A8000050X020H A8000050X030H A8000100X020H A8000100X030H A8020030X020H A8020050X020H A8020050X030H A8020100X020H A8020100X030H
VARIA8000050X020HEA 0 DKK
VARIA8000050X020H VARIA8000050X030H VARIA8000100X020H VARIA8000100X030H VARIA8020030X020H VARIA8020050X020H VARIA8020050X030H VARIA8020100X020H VARIA8020100X030H
HPLC columns, Pursuit™ UPS1,9
Kromatografikolonner
Pursuit™ UPS 1,9 columns are designed for ultimate performance on any ultra-high pressure LC system. These columns deliver sub-2 µm efficiencies when sensitivity, resolution and throughput are critical.
  • Highest efficiency particles ensure optimum resolution
  • Narrow bore columns with minimal band broadening enhance sensitivity
  • High throughput and fast separations
  • Orthogonal bonded chemistries allow rapid method development