Capillary columns, PERMABOND® CW 20 M
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
723060.10 723060.25 723060.30 723060.50 723060.60 723064.10 723296.10 723296.25 723296.30 723296.50 723296.60 723321.10 723321.25 723321.30 723321.50 723321.60 723515.10 723515.25 723517.10 723517.25 723517.30 723549.10 723549.25 723549.30 723827.10 723827.25 723827.50
MANA723060.10EA 3920 DKK
MANA723060.10 MANA723060.25 MANA723060.30 MANA723060.50 MANA723060.60 MANA723064.10 MANA723296.10 MANA723296.25 MANA723296.30 MANA723296.50 MANA723296.60 MANA723321.10 MANA723321.25 MANA723321.30 MANA723321.50 MANA723321.60 MANA723515.10 MANA723515.25 MANA723517.10 MANA723517.25 MANA723517.30 MANA723549.10 MANA723549.25 MANA723549.30 MANA723827.10 MANA723827.25 MANA723827.50
Capillary columns, PERMABOND® CW 20 M
Kromatografikolonner
Polar GC phase.
  • Suitable for solvent analysis
  • Maximum temperature for isothermal operation 220 °C (0,53 mm Int.Ø columns: 200 °C)
  • Maximum temperature for short isotherms in a temperature program 240 °C (0,53 mm Int.Ø columns: 220 °C)
Polyethylene glycol (20 kD) phase.
USP code: G16.