Chromatography columns with Rotaflo® stopcock, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CR10/20 CR10/40 CR20/30 CR20/50 CR30/50 CR60/50 CR40/50
201-0723EA 1380 DKK
201-0723 201-0725 201-0728 201-0730 201-0732 201-0738 BIBBCR40/50
Chromatography columns with Rotaflo® stopcock, Quickfit®
Kromatografikolonner
These chromatography columns are made from PYREX®, borosilicate glass.
  • With PTFE Rotaflo® stopcock for fine control of eluent flow
  • Chemically resistant glass sintered disc - porosity 0
  • Ground glass socket accepts solvent reservoirs