Capillary columns, OPTIMA WAXplus®
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
726381.30 726380.60 726383.30 726380.30 726381.60 726382.30
MANA726381.30EA 10890 DKK
MANA726381.30 MANA726380.60 MANA726383.30 MANA726380.30 MANA726381.60 MANA726382.30
Capillary columns, OPTIMA WAXplus®
Kromatografikolonner
Polar GC phase with optimized crosslinking for lower column bleeding and higher temperature stability
  • Ideal for solvent analysis and alcohols, suitable for aqueous solutions
  • Maximum temperature for isothermal operation 260 °C
  • Maximum temperature for short isotherms in a temperature program 270 °C
Polyethylene glycol phase.
USP code: G16.