HPLC columns, Polaris™ C18-A
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A2000030X030 A2000030X046 A2000050X020 A2000050X030 A2000050X046 A2000100X046 A2000150X020 A2000150X046 A2000250X046 A2001030X020 A2001030X030 A2001030X046 A2001050X020 A2001050X030 A2001050X046 A2001100X020 A2001150X020 A2001150X046 A2002250X212 A2000020X020 A2000030X020 A2000100X020 A2000100X021 A2000100X030 A2000100X212 A2000120X046 A2000125X040 A2000150X021 A2000150X030 A2000150X040 A2000150X212 A2000200X046 A2000250X020 A2000250X030 A2000250X040 A2000250X100 A2000250X212 A2000300X046 A2001020X020 A2001075X046 A2001100X030 A2001100X046 A2001150X030 A2001250X020 A2001250X030 A2001250X046 A2002250X046 A2002250X500
VARIA2000030X030EA 8320 DKK
VARIA2000030X030 VARIA2000030X046 VARIA2000050X020 VARIA2000050X030 VARIA2000050X046 VARIA2000100X046 VARIA2000150X020 VARIA2000150X046 VARIA2000250X046 VARIA2001030X020 VARIA2001030X030 VARIA2001030X046 VARIA2001050X020 VARIA2001050X030 VARIA2001050X046 VARIA2001100X020 VARIA2001150X020 VARIA2001150X046 VARIA2002250X212 VARIA2000020X020 VARIA2000030X020 VARIA2000100X020 VARIA2000100X021 VARIA2000100X030 VARIA2000100X212 VARIA2000120X046 VARIA2000125X040 VARIA2000150X021 VARIA2000150X030 VARIA2000150X040 VARIA2000150X212 VARIA2000200X046 VARIA2000250X020 VARIA2000250X030 VARIA2000250X040 VARIA2000250X100 VARIA2000250X212 VARIA2000300X046 VARIA2001020X020 VARIA2001075X046 VARIA2001100X030 VARIA2001100X046 VARIA2001150X030 VARIA2001250X020 VARIA2001250X030 VARIA2001250X046 VARIA2002250X046 VARIA2002250X500
HPLC columns, Polaris™ C18-A
Kromatografikolonner
The Polaris C18-A phase is specially designed for high-throughput and LC/MS analysis of polar analytes in drug discovery, drug metabolism, and AR&D areas.
  • Bonded with a polar group placed near the silica surface
  • Universal stationary phase with a pH range of 1,5 to 10
Provides an added retention mechanism for polar analytes. The polar group deactivates the few residual silanols through charge delocalization to enhance peak shape.