Capillary columns, OPTIMA® 17
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
726742.30 726742.50 726742.60 726744.25 726744.30 726744.50 726744.60 726747.10 726747.15 726747.25 726747.30 726755.30 726757.25 726757.30 726757.50 726757.60 726848.10 726022.15 726022.25 726022.30 726022.50 726022.60 726065.12 726065.25 726065.50 726066.25 726066.50 726067.25 726067.30 726067.50 726067.60 726351.25 726351.30 726351.50 726351.60 726742.25
MANA726742.30EA 8460 DKK
MANA726742.30 MANA726742.50 MANA726742.60 MANA726744.25 MANA726744.30 MANA726744.50 MANA726744.60 MANA726747.10 MANA726747.15 MANA726747.25 MANA726747.30 MANA726755.30 MANA726757.25 MANA726757.30 MANA726757.50 MANA726757.60 MANA726848.10 MANA726022.15 MANA726022.25 MANA726022.30 MANA726022.50 MANA726022.60 MANA726065.12 MANA726065.25 MANA726065.50 MANA726066.25 MANA726066.50 MANA726067.25 MANA726067.30 MANA726067.50 MANA726067.60 MANA726351.25 MANA726351.30 MANA726351.50 MANA726351.60 MANA726742.25
Capillary columns, OPTIMA® 17
Kromatografikolonner
Medium polar GC phase.
  • Recommended application: steroids, pesticides, drug analyses
  • Maximum temperature for isothermal operation 320 °C (0,53 mm ID columns: 300 °C)
  • Maximum temperature for short isotherms in a temperature program 340 °C (0,53 mm ID columns: 320 °C)
50% phenyl – 50% methylpolysiloxane phase.
USP code: G3.