Volumetric flasks, class B, blue markings
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FRD022P FRD024P FRD025P FRD032P FRD040P FRD026P FRD028P FRD030P FRD034P FRD036P FRD038P
612-3329EA 589 DKK
612-3329 612-3330 612-3331 612-3335 612-3336 612-3332 612-3333 612-3334 BIBBFRD034P BIBBFRD036P BIBBFRD038P
Volumetric flasks, class B, blue markings
Kolber Volumetriske kolber
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Excellent temperature resistance
Certificeringer: Complies with ISO 1042 and DIN 12664