Diamond MS syringes
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
002750 002760 002770 002780 003750 003760 003770 004750 004770 005750 005770 006750 007750 002790 003790 004790 005790
548-1584EA 858 DKK
548-1584 548-1585 548-1586 548-1587 548-1609 548-1610 548-1611 548-1622 548-1623 548-1634 548-1635 548-1648 548-1656 548-1588 548-1612 548-1624 548-1636
Diamond MS syringes
Sprøjter Sprøjter til kromatografi
Diamond MS syringes are designed to improve the sensitivity of your MS analysis.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra-smooth removable needles with choice of surface treatment
Anything in the sample flowpath can interact with the sample and affect the results. These 'Ultra Smooth' needles reduce non specific interaction between the sample and the needle.
To reduce non specific sample interaction even further, there are two needle treatment options available to avoid sample interactions due to hydrophilicity and hydrophobicity. Needles are not included with syringe and must be ordered separately.
LC needles for 10 to 50 µl syringes (manual and autosampler)
Note: Select length 57 mm for Thermo RSH applications
LC needles for 100 to 500 µl syringes (manual and autosampler)
Note: Select length 57 mm for Thermo RSH applications