1-Methyl-1,4-cyclohexadiene ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M0486-25ML
M0486-5ML
TCIAM0486-25MLEA
4450
DKK
TCIAM0486-25ML
TCIAM0486-5ML
1-Methyl-1,4-cyclohexadiene ≥95.0% (by GC)
1-Methyl-1,4-cyclohexadiene
Formel:
C₇H₁₀ Kogepunkt: 115°C Flammepunkt: 11°C |
MDL Number:
MFCD00001538 CAS-nummer: 4313-57-9 EINECS: 224-329-6 UN nummer: 3295 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...