3-Ethylcyclopentanon ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B20432.03 B20432.06 B20432.14
B20432.03EA 707 DKK
B20432.03 B20432.06 B20432.14
3-Ethylcyclopentanon ≥99%
3-Ethylcyclopentanone

Purity: 99% CAS No.: 10264-55-8 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 31°(88°F) Boiling Point: 171° UN No.: 1224 MDL No.: MFCD00045445

Formel: C₇H₁₂O
Molekylvægt: 112.17 g/mol
Kogepunkt: 171 °C
Flammepunkt: 31 °C (88 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00045445
CAS-nummer: 10264-55-8
UN nummer: 1224
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR