Methyl-4-(bromomethyl)benzoate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl 4-(bromomethyl)benzoate

B2053-10G B2053-25G
TCIAB2053-10GEA 345 DKK
TCIAB2053-10G TCIAB2053-25G
Methyl-4-(bromomethyl)benzoate ≥98.0% (by GC)
Methyl-4-(bromomethyl)benzoate
Formel: C₉H₉BrO₂
MDL Number: MFCD00032453
CAS-nummer: 2417-72-3
UN nummer: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9672da3ac89be328243d43649bea35a68848f6fb&vid=9672da3ac89be328243d43649bea35a68848f6fb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 54.0 to 57.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR