2-(Benzyloxy)benzaldehyde ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B1653-25G B1653-5G
TCIAB1653-25GEA 534 DKK
TCIAB1653-25G TCIAB1653-5G
2-(Benzyloxy)benzaldehyde ≥98.0% (by GC)
2-(Benzyloxy)benzaldehyde
Formel: C₁₄H₁₂O₂
Kogepunkt: 208°C
MDL Number: MFCD00016583
CAS-nummer: 5896-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5fbdb7adecf0bf7659bedb09b979b1d3264db5e&vid=f5fbdb7adecf0bf7659bedb09b979b1d3264db5e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR