Phenyltriethylammonium iodide ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0242-25G
P0242-5G
TCIAP0242-25GEA
2270
DKK
TCIAP0242-25G
TCIAP0242-5G
Phenyltriethylammonium iodide ≥98.0% (by HPLC, titration analysis)
Phenyltriethylammonium iodide
Formel:
C₁₂H₂₀IN |
MDL Number:
MFCD00050224 CAS-nummer: 1010-19-1 EINECS: 213-774-1 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...