9-Bromo-9-phenylfluorene ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 9-Bromo-9-phenyl-9H-fluorene

B1702-25G B1702-5G
TCIAB1702-25GEA 2220 DKK
TCIAB1702-25G TCIAB1702-5G
9-Bromo-9-phenylfluorene ≥96.0% (by GC)
9-Bromo-9-phenylfluorene
Formel: C₁₉H₁₃Br
Molekylvægt: 321.23 g/mol
MDL Number: MFCD00075522
CAS-nummer: 55135-66-5
UN nummer: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62389385f6a401fc7bec8178413d1396c58f3704&vid=62389385f6a401fc7bec8178413d1396c58f3704&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 98.0 to 102.0 deg-C
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR