4-Chromanol ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A14940.03 A14940.06
A14940.03EA 390 DKK
A14940.03 A14940.06
4-Chromanol ≥97%
4-Chromanol

Purity: 97% CAS No.: 1481-93-2 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 110°(230°F) Melting Point: 38-42° Boiling Point: 79-81°/5mm Beilstein Reference: 130800 MDL No.: MFCD00006849

Formel: C₉H₁₀O₂
Kogepunkt: 79…81 °C (5 mmHg)
Smeltepunkt: 38…42 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006849
CAS-nummer: 1481-93-2
EINECS: 216-041-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR