2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T1431-25G
T1431-5G
TCIAT1431-25GEA
3340
DKK
TCIAT1431-25G
TCIAT1431-5G
2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one ≥98.0% (by titrimetric analysis)
2,3,5,6-Tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one
Formel:
C₇H₃Br₄NO₃ Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00067043 CAS-nummer: 95111-49-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...