2-Phenylisobutyric acid
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
CAS No.: 826-55-1
Molecular Formula: C10H12O2
Formula Weight: 164.21
Physical Form: Crystalline
Melting Point: 80-82°
Boiling Point: 146.5°
Solubility: Insoluble in water
MDL No.: MFCD00014332
Formel:
C₁₀H₁₂O₂ Kogepunkt: 146,5 °C Smeltepunkt: 80…82 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00014332 CAS-nummer: 826-55-1 EINECS: 212-556-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...