2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Aminotetrafluorobenzotrifluoride

T1529-5G T1529-1G
TCIAT1529-5GEA 969 DKK
TCIAT1529-5G TCIAT1529-1G
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥97.0% (by GC)
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Formel: C₇H₂F₇N
Molekylvægt: 233.09 g/mol
Kogepunkt: 186°C
Flammepunkt: 87°C
MDL Number: MFCD00091518
CAS-nummer: 651-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10cb0334d26460891434ef12f7ab255858935c4a&vid=10cb0334d26460891434ef12f7ab255858935c4a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR