Diethyl (methylthiomethyl)phosphonate ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Diethyl ((methylthio)methyl)phosphonate
M1208-25G
M1208-5G
TCIAM1208-25GEA
1550
DKK
TCIAM1208-25G
TCIAM1208-5G
Diethyl (methylthiomethyl)phosphonate ≥97.0% (by GC)
Diethyl (methylthiomethyl)phosphonate
Formel:
C₆H₁₅O₃PS Molekylvægt: 198.220 g/mol Flammepunkt: 101°C |
MDL Number:
MFCD00015131 CAS-nummer: 28460-01-7 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...