Pentadecanenitrile 98,0 GC_ASSAY_METHOD
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Myristyl cyanide, Tetradecyl cyanide, Pentadecanonitrile
P0871-25G
P0871-5G
TCIAP0871-25GEA
2060
DKK
TCIAP0871-25G
TCIAP0871-5G
Pentadecanenitrile 98,0 GC_ASSAY_METHOD
Pentadecanenitrile
Formel:
C₁₅H₂₉N Molekylvægt: 223.397 g/mol |
MDL Number:
MFCD00019849 CAS-nummer: 18300-91-9 EINECS: 242-184-7 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...