Thuliumstandard, 1g/l Tm i salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86719.180EA 190.4 DKK
86719.180 86719.260
Thuliumstandard, 1g/l Tm i salpetersyre AVS TITRINORM standard til AAS
Standarder Kationiske standarder Thulium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%